3-Amino-6-bromopyrazine-2-carbonitrile 98%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
A106784-1G
A106784-250MG
A106784-5G
A106784-25G
A106784-10G
77008-648EA
18
USD
77008-648
77008-650
77610-844
77682-982
77889-741
3-Amino-6-bromopyrazine-2-carbonitrile 98%, Ambeed.Inc
3-Amino-6-bromopyrazine-2-carbonitrile
Caution: For research use only.
Formula:
C₅H₃BrN₄ MW: 199.02 g/mol |
MDL Number:
MFCD06658300 CAS Number: 17231-51-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...