Tricine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid, Tricine buffer, N-(Tris(hydroxymethyl)methyl)glycine
A102157-100G
A102157-25G
A102157-1000G
A102157-500G
A102157-10G
77025-198EA
33
USD
77025-198
77025-200
77548-926
77548-928
77599-496
Tricine 98%
Tricine
Caution: for Research Use Only
Formula:
(HOCH₂)₃CNHCH₂CO₂H MW: 179.17 g/mol Melting Pt: 187 |
MDL Number:
MFCD00004277 CAS Number: 5704-04-1 Merck Index: 13,09724 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...