Sodium chloride solution 1 M, in single-use containers, HyClone™ for bioprocessing
Supplier: Cytiva
HyClone™ NaCl solutions in single-use bags offer a convenient format for many upstream and downstream bioprocessing applications.
- Manufactured in ISO 9001 and ISO 13485 certified facility.
- Lot-specific certificate of analysis (CoA) for each lot produced.
- Intended for further manufacturing or research use. Not for diagnostic or therapeutic use.
- 0.2 µm sterile filtered.
- Provided in single-use bag containers.
- Raw material is EP, USP, and JP grade.
These products are manufactured upon order (100 L minimum order quantity). HyClone™ NaCl in single-use bags is well-suited for use with: Cytiva's chromatography resins.
Available in 1 M and 2 M sodium chloride concentrations up to 200 L. Contact us for information around bag configurations or custom bulk formulations.
Formula:
NaCl Storage Temperature: Ambient |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 |
Specification Test Results
Storage | Room Temperature as defined by regional Pharmacopeia (15-30°C in US; 15-25°C in EU) |
Endotoxin Concentration | ≤ 1 EU/mL |
Certifications/Compliance | Manufactured in a ISO9001 and ISO13485 certified facility |
Shelf life | 12 Months |
For use with | Chromatography resins, IC systems, ÄKTA hardware systems |
Appearance | Clear, colorless solution |
Sterility | No Growth |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...