NOVAX® Class 0 Insulating Gloves with Straight Cuff, Orange
Supplier: PIP
147-0-11/10 147-0-11/10.5 147-0-11/11 147-0-11/12 147-0-11/6 147-0-11/7 147-0-11/8 147-0-11/8.5 147-0-11/9 147-0-11/9.5
76944-310PR 70.28 USD
76944-310 76944-312 76944-314 76944-316 76944-298 76944-300 76944-302 76944-304 76944-306 76944-308
NOVAX® Class 0 Insulating Gloves with Straight Cuff, Orange
Gloves
Rubber insulating gloves are critical PPE for EV manufacturing. They help protect automotive workers against electrical shock if contact is made to an energized surface.
  • Strong, flexible, durable, and dielectric
Critical PPE for EV manufacturing. Gloves should meet and/or exceed the requirements of the ASTM D120-14a Standard specification for rubber insulating gloves. Gloves should also be electrically tested following ASTM D120-14a.
Certifications: Gloves should meet and/or exceed the requirements of the ASTM D120-14a.