NOVAX® Class 00 Electrical Safety Kits, Orange Rubber Insulating Gloves
Supplier: PIP
147-SK-00/10-KIT 147-SK-00/11-KIT 147-SK-00/12-KIT 147-SK-00/7-KIT 147-SK-00/8-KIT 147-SK-00/9-KIT
76944-348EA 109.36 USD
76944-348 76944-350 76944-352 76944-342 76944-344 76944-346
NOVAX® Class 00 Electrical Safety Kits, Orange Rubber Insulating Gloves
Gloves
Electrical Safety Kits allow users to pick the class that an application requires, the color and size rubber gloves required, and Protective Industrial Products does the rest by assembling a kit that works.
  • 1 pair of rubber insulating gloves
  • 1 pair of leather protector gloves
  • 11" nylon storage bag for gloves
Critical PPE for EV manufacturing. Gloves should meet and/or exceed the requirements of the ASTM D120-14a Standard Specification for Rubber Insulating gloves. Gloves should also be electrically tested following ASTM D120-14a.
Certifications: Gloves should meet and/or exceed the requirements of the ASTM D120-14a.