Globe Glass™ Erlenmeyer Flasks, Heavy Duty
Supplier: Globe Scientific
8430250 8430500 8431000 8432000 8434000
76683-714BX 137.48 USD
76683-714 76683-734 76683-690 76683-724 76683-682
Globe Glass™ Erlenmeyer Flasks, Heavy Duty
Flasks Erlenmeyer Flasks
Globe Glass™ heavy duty Erlenmeyer flasks meet ASTM E1406 specifications.
  • Made exclusively from ASTM E438, Type I, Class A, 3.3 borosilicate glass
  • GlassGuard™ packaging system protects against transit damage
  • Thicker walls protect against damage
  • Dual metric scale graduations
  • Durable white enamel printing
  • Large white marking area
Thick, heavy-duty walls improve mechanical strength and allow use in filtering applications not requiring side tubulation. Globe Glass heavy duty Erlenmeyer flasks are designed with reinforced, beaded rims for strength and resistance to chipping.
Certifications: Meet ASTM E1406, Type I Specifications.