Globe Glass™ Reusable Serological Pipettes
Supplier: Globe Scientific
8500005 8500010 8500025 8510000 8510000A 8510001 8510001A 8510002 8510005 8510010 8500001A 8500002 8510025
76683-726BX 84.94 USD
76683-726 76683-698 76683-676 76683-730 76683-686 76683-688 76683-694 76683-708 76683-678 76683-666 76683-674 76683-706 76683-680
Globe Glass™ Reusable Serological Pipettes
Pipettes Serological Pipettes
Globe Glass™ serological pipettes measure and dispense specific volumes of liquid and are calibrated to ASTM E1044 standards.
  • Meet ASTM E1044 Style I specifications
  • Made of ASTM E438, Type I, Class B, 5.1 borosilicate glass
  • GlassGuard™ packaging system protects against transit damage
  • Calibrated TD – 'To Deliver'
  • Class A calibration tolerance for highest accuracy and the most critical laboratory work
  • Class B calibration tolerance for general laboratory work
Globe Glass reusable serological pipettes are calibrated TD 'To Deliver' and must be blown outMore Product Information