Globe Glass™ Square Media Bottles
Supplier: Globe Scientific
8120500 8121000 8122000
76683-702BX 227.13 USD
76683-702 76683-700 76683-722
Globe Glass™ Square Media Bottles
Bottles Glass Bottles
Globe Glass™ square, wide mouth media bottles are designed with an extra-wide mouth for ease of filling, dispensing, and pipetting.
  • ASTM E438, Type I, Class A, borosilicate 3.3 glass
  • GlassGuard™ packaging system protects against transit damage
  • Pouring ring for drip-free liquid transfer
  • High resistance to chemicals
  • Single scale metric graduations
  • Durable white enamel printing
  • Large white marking area
The square shape improves storage and transport efficiency. A drip-free pour ring and autoclavable polypropylene (PP) screw cap enable sterilization of media and reagents. Globe Glass™ square wide mouth media bottles are available in sizes 500, 1000 and 2000 ml.