Laser Cassette Printers, VEGA/NOVA
Supplier: Epredia
8000039 8000038 NOVA05-FT-PCDI VEGA05-FT-PCDI
76540-418EA 38282.28 USD
76540-418 76540-420 76540-446 76540-444
Laser Cassette Printers, VEGA/NOVA
Printers Cassette and Slide Printers
The Epredia Laser Cassette Printing technology produces high quality print combined with a patented chemical additive in the cassette to deliver consistency and efficiency. Built in interface connectivity ensures LIS/LIM compatibility.
  • Epredia NOVA laser Printer is a single cassette hopper printer providing an economical option for laboratories
  • Three additional hoppers are included for convenient cassette exchanges
  • Epredia VEGA laser printer has six cassette hoppers and supports laboratories printing a variety of cassette colors and styles
  • Laser Technology eliminates ancillary components such as print heads & ribbons and simplifies operation
  • High resolution printing, 600 dpi, improves printing sharpness and clarity of characters allowing more characters to be embedded on cassette surface
  • Fully automated system with fast print speed, five seconds per cassette, allows the user to quickly and easily print cassettes needed for a case at accessioning or grossing
  • Laser system with cassette additive print technology ensures permanent and delible marking that are extremely durable overtime
  • Small footprint reduces the concern of space constraint in the lab
  • Quiet printing reduces distraction during grossing and accessioning
Workflow Efficiencies –
Epredia laser printers are designed to create crisp, accurate and indelible print quality for laboratories concerned about workflow bottlenecks caused by the need for reprinting cassettes with low-quality or unreadable printing at accessioning.
High Printing Quality –
•Epredia laser printers create sustainable and indelible printed information where patient data can be identified accurately throughout the lifetime of the cassette supporting laboratories and patients.
•Character Types: Alphanumeric.
•Barcode Types: Linear, Bidimensional, ASCII Codes.
High Throughput –
Epredia laser printers incorporate a highly efficient technology helping labs with higher printing demands utilize the laser printing capabilities and automated cassette feeder feature to achieve high throughput and faster turn-around times.
Low Operations Cost –
Epredia laser printers do not require replacement of ancillary components significantly decreasing operation costs and lowers any intensive maintenance helping lab managers from being burdened with high operation costs.