Acesulfame potassium 99%

Supplier: Ambeed
Warning

A334842-25G A334842-100G A334842-1000G A334842-500G
76978-312EA 9 USD
76978-312 77612-922 77672-712 77672-714
Acesulfame potassium 99%
Acesulfame potassium

Caution: for Research Use Only

Formula: C₄H₄KNO₄S
MDL Number: MFCD00043833
CAS Number: 55589-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR