Toluene-α-sulfonyl fluoride 95%

Supplier: Ambeed
Danger

A767839-100MG A767839-1G A767839-5G A767839-10G
76981-474EA 5 USD
76981-474 77552-368 77614-362 77674-110
Toluene-α-sulfonyl fluoride 95%
Toluene-α-sulphonyl fluoride

Caution: for Research Use Only

Formula: C₇H₇FO₂S
MDL Number: MFCD00007424
CAS Number: 329-98-6
UN: 2928
ADR: 6.1,8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR