1-(3-Aminopropyl)imidazole 97%

Supplier: Ambeed
Danger

Synonyms: N-(3-Aminopropyl)-imidazole, Imidazole-1-propylamine, 1-(3-Aminopropyl)-1H-imidazole

A342606-1G A342606-25G A342606-5G
76978-424EA 5 USD
76978-424 76978-426 76978-428
1-(3-Aminopropyl)imidazole 97%
1-(3-Aminopropyl)imidazole

Caution: for Research Use Only

Formula: C₆H₁₁N₃
MW: 125.17 g/mol
MDL Number: MFCD00009819
CAS Number: 5036-48-6
UN: 3267
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR