4-Amino-5-methoxy-6-chloropyrimidine 95%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-6-chloro-5-methoxypyrimidine

A241068-1G A241068-250MG A241068-25G A241068-5G
76976-448EA 32 USD
76976-448 76976-450 76976-452 76976-454
4-Amino-5-methoxy-6-chloropyrimidine 95%
4-Amino-5-methoxy-6-chloropyrimidine

Caution: for Research Use Only

Formula: C₅H₆ClN₃O
MW: 159.574 g/mol
MDL Number: MFCD07787635
CAS Number: 5018-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR