4-(Trifluoromethylthio)phenol 98%

Supplier: Ambeed
Warning

Synonyms: 4-[(Trifluoromethyl)sulphanyl]phenol4-[(Trifluoromethyl)sulfanyl]phenol

A156506-5G A156506-1G A156506-10G A156506-25G
76973-338EA 13.8 USD
76973-338 76973-334 76973-336 76973-340
4-(Trifluoromethylthio)phenol 98%
4-(Trifluoromethylthio)phenol

Caution: for Research Use Only

Formula: C₇H₅F₃OS
MW: 194.18 g/mol
MDL Number: MFCD00036035
CAS Number: 461-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR