3-Methoxy-5-(trifluoromethyl)phenol 97%

Supplier: Ambeed
Warning

Synonyms: 3-hydroxy-5-methoxy-benzotrifluoride

A148678-1G A148678-250MG A148678-5G
76972-996EA 29 USD
76972-996 76972-998 76973-000
3-Methoxy-5-(trifluoromethyl)phenol 97%
3-Methoxy-5-(trifluoromethyl)phenol

Caution: for Research Use Only

Formula: C₈H₇F₃O₂
MDL Number: MFCD09025418
CAS Number: 349-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR