tert-Butyl-N-[2-hydroxy-1,1-bis(hydroxymethyl)ethyl]carbamate 95%

Supplier: Ambeed
Warning

A703059-100MG A703059-1G A703059-250MG A703059-25G A703059-5G
76981-108EA 5 USD
76981-108 76981-110 76981-112 76981-114 76981-116
tert-Butyl-N-[2-hydroxy-1,1-bis(hydroxymethyl)ethyl]carbamate 95%
tert-Butyl-N-[2-hydroxy-1,1-bis(hydroxymethyl)ethyl]carbamate

Caution: for Research Use Only

Formula: C₉H₁₉NO₅
MDL Number: MFCD14581676
CAS Number: 146651-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR