5-Amino-2-chloro-3-methylpyridine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Amino-2-chloro-3-picoline, 6-Chloro-5-methylpyridin-3-amine
A127047-100G
A127047-10G
A127047-1G
A127047-25G
A127047-5G
76971-908EA
171
USD
76971-908
76971-910
76971-912
76971-914
77670-792
5-Amino-2-chloro-3-methylpyridine 97%
5-Amino-2-chloro-3-methylpyridine
Caution: for Research Use Only
Formula:
C₆H₇ClN₂ |
MDL Number:
MFCD03095087 CAS Number: 38186-82-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...