1,3-Diamino-2-propanol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,3-Diaminopropan-2-ol
, 1,3-Diamino-2-hydroxypropane
A206041-500G
A206041-1000G
A206041-25G
A206041-100G
A206041-10G
A206041-5G
A206041-1G
76975-358EA
6.18
USD
76975-358
76975-346
76975-348
76975-350
76975-352
76975-354
76975-356
1,3-Diamino-2-propanol 98%
1,3-Diamino-2-propanol
Caution: for Research Use Only
Formula:
C₃H₁₀N₂O MW: 90.13 g/mol |
MDL Number:
MFCD00008142 CAS Number: 616-29-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...