N-Boc-β-phenyl-D-phenylalanine 97%

Supplier: Ambeed
Warning

A187365-1G A187365-250MG A187365-5G A187365-10G
76974-700EA 23 USD
76974-700 76974-702 76974-704 77881-401
N-Boc-β-phenyl-D-phenylalanine 97%
N-Boc-beta-phenyl-D-phenylalanine

Caution: for Research Use Only

Formula: C₂₀H₂₃NO₄
MDL Number: MFCD00191187
CAS Number: 143060-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR