1-Hydroxy-1,2-benziodoxol-3-one 97%

Supplier: Ambeed
Danger

Synonyms: 1-Hydroxy-3-oxo-1,3-dihydro-1,2-benzoiodoxole, IBA

A604849-1G A604849-250MG A604849-5G A604849-10G A604849-25G
76980-526EA 14 USD
76980-526 76980-528 76980-530 77673-632 77673-634
1-Hydroxy-1,2-benziodoxol-3-one 97%
1-Hydroxy-1,2-benziodoxol-3-one

Caution: for Research Use Only

Formula: C₇H₅IO₃
MW: 264.1 g/mol
MDL Number: MFCD01658875
CAS Number: 131-62-4
UN: 1479
ADR: 5.1 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR