Hippuric acid 95%

Supplier: Ambeed
Warning

A216626-100G A216626-25G A216626-500G
76975-704EA 23 USD
76975-704 76975-706 76975-708
Hippuric acid 95%
Hippuric acid

Caution: for Research Use Only

Formula: C₆H₅C(O)NHCH₂COOH
MDL Number: MFCD00002692
CAS Number: 495-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR