7-Amino-3-Cephem-4-Carboxylic Acid 97%

Supplier: Ambeed
Warning

Synonyms: 7-Anca, (6R,7R)-7-Amino-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-Amino-3-cephem-4-carboxylic acid

A343958-1G A343958-5G
76978-436EA 14 USD
76978-436 76978-438
7-Amino-3-Cephem-4-Carboxylic Acid 97%
7-Amino-3-Cephem-4-Carboxylic Acid

Caution: for Research Use Only

Formula: C₇H₈N₂O₃S
MDL Number: MFCD08063340
CAS Number: 36923-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR