Backup Systems for ULT Freezers
Supplier: Thermo Fisher Scientific
Thermo Scientific™
332353G06 332353G05 332353G02 332353G01
76779-132EA 3570 USD
76779-132 76779-134 76779-130 76779-136
Backup Systems for ULT Freezers
Freezers Ultra Low Temperature Freezers
CO₂ and LN₂ backup system options for ultra-low temperature chest freezers.
  • Factory installed, operates on independent backup probe
Sample security is paramount in cold storage and backup systems help ensure continued protection in the event of a refrigeration system issue with cold storage equipment. LN₂ and CO₂ backup options help maintain temperatures below −60 °C until a unit's primary refrigeration system returns to operation.
Ordering information: Add to ordering configuration with ULT chest freezer.