Methyl 4-(methoxymethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

36257-1G 36257-25G 36257-5G
76941-438EA 117.76 USD
76941-438 76941-440 76941-442
Methyl 4-(methoxymethyl)benzoate ≥97%
Methyl 4-(methoxymethyl)benzoate

Caution: for Research Use Only

Formula: C₁₀H₁₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD10690281
CAS Number: 1719-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR