Methyl 2,6-dichloro-4-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

26992-1G 26992-25G 26992-500MG 26992-5G
76939-004EA 275.84 USD
76939-004 76939-006 76939-008 76939-010
Methyl 2,6-dichloro-4-fluorobenzoate ≥97%
Methyl 2,6-dichloro-4-fluorobenzoate

Caution: for Research Use Only

Formula: C₈H₅Cl₂FO₂
Storage Temperature: Refrigerator
MDL Number: MFCD23135661
CAS Number: 232275-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR