6-(2-Methylpropoxy)pyridine-3-boronic acid ≥97%

Supplier: AOBChem USA

Warning

23993-1G 23993-25G 23993-500MG 23993-5G
76938-368EA 391.48 USD
76938-368 76938-370 76938-372 76938-374
6-(2-Methylpropoxy)pyridine-3-boronic acid ≥97%
6-(2-Methylpropoxy)pyridine-3-boronic acid

Caution: for Research Use Only

Formula: C₉H₁₄BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD24368719
CAS Number: 2406206-23-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR