N-(2-Diethylaminoethyl)-1-naphthylamine oxalate ≥95% research grade
Supplier: AOBChem USA
|
Warning
|
Synonyms:
Tsuda's Reagent
, N,N-Diethyl-N'-1-naphthylethylenediamine Oxalate
, N1,N1-Diethyl-N2-(naphthalen-1-yl)ethane-1,2-diamine oxalate
10097-25G
76936-336EA
129.78
USD
76936-336
N-(2-Diethylaminoethyl)-1-naphthylamine oxalate ≥95% research grade
N-(2-Diethylaminoethyl)-1-naphthylamine oxalate
Formula:
C₁₆H₂₄N₂O₄ MW: 332.40 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00060156 CAS Number: 29473-53-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...