5-(tert-Butyl)-2-chlorobenzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
52448-1G
52448-25G
52448-500MG
52448-5G
76933-134EA
186.72
USD
76933-134
76933-136
76933-138
76933-140
5-(tert-Butyl)-2-chlorobenzaldehyde ≥97%
5-(tert-Butyl)-2-chlorobenzaldehyde
Caution: for Research Use Only
Formula:
C₁₁H₁₃ClO Storage Temperature: Refrigerator |
CAS Number:
1160574-90-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...