4-Chloro-2-fluoro-3-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

32191-25G 32191-5G
76930-852EA 595.17 USD
76930-852 76930-854
4-Chloro-2-fluoro-3-methoxybenzoic acid ≥97%
4-Chloro-2-fluoro-3-methoxybenzoic acid

Caution: for Research Use Only

Formula: C₈H₆ClFO₃
Storage Temperature: Ambient
MDL Number: MFCD19687204
CAS Number: 1169870-80-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR