5-Bromo-3-chloro-2,6-dimethoxypyridine ≥97%

Supplier: AOBChem USA

Warning

30744-1G 30744-25G 30744-500MG 30744-5G
76930-368EA 196.27 USD
76930-368 76930-370 76930-372 76930-374
5-Bromo-3-chloro-2,6-dimethoxypyridine ≥97%
5-Bromo-3-chloro-2,6-dimethoxypyridine

Caution: for Research Use Only

Formula: C₇H₇BrClNO₂
Storage Temperature: Ambient
MDL Number: MFCD29915587
CAS Number: 1917346-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR