4-Bromo-3-chloro-2-fluorobenzyl alcohol ≥97% research grade

Supplier: AOBChem USA

Warning

27504-1G 27504-25G 27504-500MG 27504-5G
76929-510EA 343.74 USD
76929-510 76929-512 76929-514 76929-516
4-Bromo-3-chloro-2-fluorobenzyl alcohol ≥97% research grade
4-Bromo-3-chloro-2-fluorobenzyl alcohol
Formula: C₇H₅BrClFO
Storage Temperature: Refrigerator
MDL Number: MFCD28142598
CAS Number: 1702154-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR