2-Amino-5-bromo-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

75733-25G 75733-5G
76905-888EA 375.56 USD
76905-888 76905-890
2-Amino-5-bromo-3-methylbenzoic acid ≥97%
2-Amino-5-bromo-3-methylbenzoic acid

Caution: for Research Use Only

Formula: C₈H₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD05664860
CAS Number: 206548-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR