2-Chloro-5-fluoro-4-methylbenzoic acid ≥95%
Supplier: AOBChem USA
|
Warning
|
49250-1G
49250-25G
49250-5G
76904-452EA
62.6
USD
76904-452
76904-454
76904-456
2-Chloro-5-fluoro-4-methylbenzoic acid ≥95%
2-Chloro-5-fluoro-4-methylbenzoic acid
Caution: for Research Use Only
Formula:
C₈H₆ClFO₂ Storage Temperature: Ambient |
CAS Number:
103877-61-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...