2,5-Dichloro-3-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

35004-1G 35004-25G 35004-500MG 35004-5G
76903-070EA 203.7 USD
76903-070 76903-072 76903-074 76903-076
2,5-Dichloro-3-(trifluoromethyl)benzoic acid ≥97%
2,5-Dichloro-3-(trifluoromethyl)benzoic acid

Caution: for Research Use Only

Formula: C₈H₃Cl₂F₃O₂
MDL Number: MFCD27939555
CAS Number: 871254-76-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR