5-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Formyl-4-methylphenylboronic acid pinacol ester

33528-1G 33528-25G 33528-500MG 33528-5G
76902-592EA 223.85 USD
76902-592 76902-594 76902-596 76902-598
5-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde ≥97% research grade
5-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Formula: C₁₄H₁₉BO₃
Storage Temperature: Ambient
MDL Number: MFCD13181634
CAS Number: 1418129-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR