4-Hydroxy-2,6-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Dimethyl-4-hydroxybenzaldehyde

26300-1G 26300-25G 26300-500MG 26300-5G
76900-338EA 381.93 USD
76900-338 76900-340 76900-342 76900-344
4-Hydroxy-2,6-dimethylbenzaldehyde ≥97%
4-Hydroxy-2,6-dimethylbenzaldehyde

Caution: for Research Use Only

Formula: C₉H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00016979
CAS Number: 70547-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR