2-Fluoro-3-methoxypyridine-4-boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: (2-Fluoro-3-methoxypyridin-4-yl)boronic acid

24733-1G 24733-25G 24733-500MG 24733-5G
76899-628EA 356.47 USD
76899-628 76899-630 76899-632 76899-634
2-Fluoro-3-methoxypyridine-4-boronic acid ≥97% research grade
2-Fluoro-3-methoxypyridine-4-boronic acid
Formula: C₆H₇BFNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD16611242
CAS Number: 1451392-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR