3-(5-Bromo-pyridin-3-yl)benzaldehyde ≥95% research grade

Supplier: AOBChem USA

Warning

35125-1G 35125-25G 35125-500MG 35125-5G
76915-346EA 302.36 USD
76915-346 76915-348 76915-350 76915-352
3-(5-Bromo-pyridin-3-yl)benzaldehyde ≥95% research grade
3-(5-Bromo-pyridin-3-yl)benzaldehyde
Formula: C₁₂H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD28337624
CAS Number: 1171896-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR