3-Chlor-2-methoxy-4-pyridineboronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
3-Chloro-2-methoxypyridine-4-boronic acid pinacol ester
32602-1G
32602-25G
32602-5G
76914-312EA
112.46
USD
76914-312
76914-314
76914-316
3-Chlor-2-methoxy-4-pyridineboronic acid pinacol ester ≥97%
3-Chlor-2-methoxy-4-pyridineboronic acid pinacol ester
Caution: for Research Use Only
Formula:
C₁₂H₁₇BClNO₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD06798258 CAS Number: 1073353-73-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...