3,5-Dichloro-4-isopropoxyphenylboronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
3,5-Dichloro-4-isopropoxyphenyboronic acid
19896-1G
19896-25G
19896-5G
76909-858EA
94.43
USD
76909-858
76909-860
76909-862
3,5-Dichloro-4-isopropoxyphenylboronic acid ≥97%
3,5-Dichloro-4-isopropoxyphenylboronic acid
Caution: for Research Use Only
Formula:
C₉H₁₁BCl₂O₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD15143466 CAS Number: 1218790-62-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...