(m-Acrylamidophenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Acrylamidophenylboronic acid, (3-Acrylamidophenyl)boronic acid

13425-25G 13425-5G
76907-166EA 495.45 USD
76907-166 76907-168
(m-Acrylamidophenyl)boronic acid ≥97% research grade
(m-Acrylamidophenyl)boronic acid
Formula: C₉H₁₀BNO₃
MW: 191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09025755
CAS Number: 99349-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR