3-Iodophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

12111-25G 12111-5G
76906-902EA 201.58 USD
76906-902 76906-904
3-Iodophenylboronic acid ≥97%
3-Iodophenylboronic acid

Caution: for Research Use Only

Formula: C₆H₆BIO₂
Storage Temperature: Ambient
MDL Number: MFCD01319015
CAS Number: 221037-98-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR