2-(5-Bromo-2,3,4-trifluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%

Supplier: AOBChem USA

Warning

36498-1G 36498-25G 36498-500MG 36498-5G
76888-458EA 274.78 USD
76888-458 76888-460 76888-462 76888-464
2-(5-Bromo-2,3,4-trifluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(5-Bromo-2,3,4-trifluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Caution: for Research Use Only

Formula: C₁₂H₁₃BBrF₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD12405350
CAS Number: 1073339-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR