2-(2-Fluorophenyl)-1,3-dioxolane ≥97% research grade

Supplier: AOBChem USA

Warning

27070-25G 27070-5G
76886-122EA 360.71 USD
76886-122 76886-124
2-(2-Fluorophenyl)-1,3-dioxolane ≥97% research grade
2-(2-Fluorophenyl)-1,3-dioxolane
Formula: C₉H₉FO₂
Storage Temperature: Refrigerator
MDL Number: MFCD06208905
CAS Number: 133393-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR