Ethyl 2-chloro-5-fluoroisonicotinate ≥97% research grade

Supplier: AOBChem USA

Warning

83770-1G 83770-25G 83770-500MG 83770-5G
76944-058EA 168.69 USD
76944-058 76944-060 76944-062 76944-064
Ethyl 2-chloro-5-fluoroisonicotinate ≥97% research grade
Ethyl 2-chloro-5-fluoroisonicotinate
Formula: C₈H₇ClFNO₂
Storage Temperature: Ambient
CAS Number: 1214346-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR