Methyl 2-chloro-6-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

63221-1G 63221-25G 63221-5G
76943-706EA 96.55 USD
76943-706 76943-708 76943-710
Methyl 2-chloro-6-methoxybenzoate ≥97%
Methyl 2-chloro-6-methoxybenzoate

Caution: for Research Use Only

Formula: C₉H₉ClO₃
Storage Temperature: Ambient
MDL Number: MFCD18398978
CAS Number: 936479-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR