6-Chloro-5-iodonicotinic acid ≥97%

Supplier: AOBChem USA

Warning

38544-25G 38544-5G
76942-252EA 481.65 USD
76942-252 76942-254
6-Chloro-5-iodonicotinic acid ≥97%
6-Chloro-5-iodonicotinic acid

Caution: for Research Use Only

Formula: C₆H₃ClINO₂
Storage Temperature: Ambient
MDL Number: MFCD11100073
CAS Number: 59782-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR