Methyl 3-(benzyloxy)-2-fluoro-5-(trifluoromethyl)benzoate ≥95% research grade

Supplier: AOBChem USA

Warning

38087-1G 38087-25G 38087-500MG 38087-5G
76942-060EA 236.59 USD
76942-060 76942-062 76942-064 76942-066
Methyl 3-(benzyloxy)-2-fluoro-5-(trifluoromethyl)benzoate ≥95% research grade
Methyl 3-(benzyloxy)-2-fluoro-5-(trifluoromethyl)benzoate
Formula: C₁₆H₁₂F₄O₃
Storage Temperature: Refrigerator
MDL Number: MFCD32632304
CAS Number: 2404734-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR