6-Bromo-1-chloro-2-naphthol ≥97%

Supplier: AOBChem USA

Warning

20214-1G 20214-25G 20214-500MG 20214-5G
76937-606EA 283.27 USD
76937-606 76937-608 76937-610 76937-612
6-Bromo-1-chloro-2-naphthol ≥97%
6-Bromo-1-chloro-2-naphthol

Caution: for Research Use Only

Formula: C₁₀H₆BrClO
Storage Temperature: Refrigerator
MDL Number: MFCD11109963
CAS Number: 102169-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR