5-Chloro-2-methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 5-Chloro-2-methoxypyridine-3-boronic acid pinacol ester

21322-1G 21322-25G 21322-500MG 21322-5G
76927-412EA 162.32 USD
76927-412 76927-414 76927-416 76927-418
5-Chloro-2-methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥97% research grade
5-Chloro-2-methoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Formula: C₁₂H₁₇BClNO₃
Storage Temperature: Ambient
MDL Number: MFCD09878898
CAS Number: 1083168-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR